Please login to your online account to display your discounted pricing

OPTIZYME™ HindIII, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80065

 View more versions of this product

Catalog No. BP8006-5

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Keep container tightly closed
Quantity 15000U
Storage Buffer 10mM Tris HCl (pH 7.5 at 25°C), 250mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
Product Name HindIII
Format Liquid
Includes 25 to 50% Water, >50% Glycerin, 1 to 2.5% Potassium Chloride
Applications >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with HaeIII
Units 15,000U
Concentration 10U/μL
Cut Site A.AGCTT
For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with HaeIII
Incubator Temperature 37°C
Format Liquid
Product Name HindIII
Quantity 15000U