Please login to your online account to display your discounted pricing

OPTIZYME™ HaeIII (BsuRI), Fisher BioReagents™


Manufacturer: fisher scientific company  BP80025

 View more versions of this product

Catalog No. BP8002-5

  • / Each
 

Description & Specifications

Specifications

Storage Buffer 10mM Tris HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
Incubator Temperature 37°C
Quantity 10000U
For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with HaeIII
Format Liquid
Product Name HaeIII (BsuRI)
Includes 25 to 50% Water, >50% Glycerin
Units 10,000U
Storage Requirements Keep container tightly closed
Product Name HaeIII (BsuRI)
Concentration 10U/μL
Cut Site GG.CC
Quantity 10000U
Applications >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with HaeIII
Format Liquid