Please login to your online account to display your discounted pricing

OPTIZYME™ EcoRI, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80035

 View more versions of this product

Catalog No. BP8003-5

  • / Each
 

Description & Specifications

Specifications

Incubator Temperature 37°C
Cut Site G.AATTC
Storage Buffer 10mM Potassium Phosphate (pH 7.4 at 25°C), 300mM NaCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 0.15% Triton™ X-100, 50% Glycerol
For Use With (Application) >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with EcoRI
Storage Requirements Keep container tightly closed
Includes 25 to 50% Water, >50% Glycerin, 2.5 to10% Sodium Chloride
Format Liquid
Product Name EcoRI
Format Liquid
Applications >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with EcoRI
Concentration 10U/μL
Product Name EcoRI
pH 7.4
Quantity 15,000U
Quantity 15,000U