Please login to your online account to display your discounted pricing

OPTIZYME™ DdeI (HpyF31), Fisher BioReagents™


Manufacturer: fisher scientific company  BP80601

 View more versions of this product

Catalog No. BP8060-1

  • / Each
 

Description & Specifications

Specifications

Product Name Dde I
Incubator Temperature 37°C
pH 7.4
Includes 25 to 50% Water, >50% Glycerin
Quantity 500U
For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digesiton with DdeI
Storage Requirements Keep container tightly closed
Format Liquid
Cut Site C.TNAG
Applications >95% of DNA fragments can be ligated and re-cut after 50-fold over-digesiton with DdeI
Format Liquid
Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM DTT, 1mM EDTA, 0.2mg/mL BSA, 50% Glycerol
Concentration 10U/μL
Product Name Dde I
Quantity 500U