Please login to your online account to display your discounted pricing

OPTIZYME™ ClaI (Bsu15I), Fisher BioReagents™


Manufacturer: fisher scientific company  BP80245

 View more versions of this product

Catalog No. BP8024-5

  • / Each
 

Description & Specifications

Specifications

Product Name ClaI (Bsu15I)
Storage Buffer 10mM Tris HCl (pH 8 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA, 50% Glycerol
Storage Requirements Keep container tightly closed
Format Liquid
Concentration 10U/μL
pH 7.4
Incubator Temperature 37°C
Includes >50% Water, 25 to 50% Glycerin
Quantity 2500U
For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with ClaI
Cut Site AT.CGAT
Product Name ClaI (Bsu15I)
Quantity 2500U
Applications >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with ClaI
Format Liquid