Please login to your online account to display your discounted pricing

OPTIZYME™ BglII, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80141

 View more versions of this product

Catalog No. BP8014-1

  • / Each
 

Description & Specifications

Specifications

Product Name BglII
Incubator Temperature 37°C
pH 7.4
Includes 25 to 50% Water, >50% Glycerin, 1 to 2.5% Sodium Chloride
Storage Requirements Keep container tightly closed
Format Liquid
Concentration 10U/μL
Cut Site A.GATCT
Quantity 1000U
For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with BglII
Storage Buffer 10mM Tris HCl (pH 8.2 at 25°C), 200mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/mL BSA, 50% Glycerol
Product Name BglII
Applications >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with BglII
Quantity 1000U
Format Liquid