Please login to your online account to display your discounted pricing

OPTIZYME™ BamHI, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80051

 View more versions of this product

Catalog No. BP8005-1

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with BamHI
Cut Site G.GATCC
pH 7.4
Incubator Temperature 37°C
Format Liquid
Product Name BamHI
Concentration 10U/μL
Quantity 5000U
Format Liquid
Storage Requirements Keep container tightly closed
Includes 25 to 50% Water, >50% Glycerin, 1 to 2.5% Sodium Chloride
Storage Buffer 10mM Tris HCl (pH 7.4 at 25°C), 200mM NaCl, 1mM DTT, 0.1mM EDTA, 0.15% Triton™ X-100, 0.2mg/mL BSA, 50% Glycerol
Applications >95% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with BamHI
Product Name BamHI
Quantity 5000U