Please login to your online account to display your discounted pricing

OPTIZYME™ Alkaline Phosphatase, Fisher BioReagents™

$170.63 - $434.22

Specifications

Color Colorless
Concentration 1U/μL
Product Name Alkaline Phosphatase
Format Solid
For Use With (Application) DePhosphorylates cloning vector DNA (10 min. at 37°C), fast thermo-inactivation (5 min. at 75°C), Nucleotide degradation prior to sequencing PCR products, active on Nucleotides, Protein dephosphorylation, restriction En
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

BP80971

 
fisher scientific company
BP80971
1000U Each for $170.63

BP80975

 
fisher scientific company
BP80975
3000U Each for $434.22
Description & Specifications

Specifications

Color Colorless
Concentration 1U/μL
Product Name Alkaline Phosphatase
Format Solid
For Use With (Application) DePhosphorylates cloning vector DNA (10 min. at 37°C), fast thermo-inactivation (5 min. at 75°C), Nucleotide degradation prior to sequencing PCR products, active on Nucleotides, Protein dephosphorylation, restriction En
pH 7.4
Source E. coli cells with a cloned bacterial Alkaline Phosphatase gene
Storage Buffer OPTIZYME™ AP Buffer 10X : Tris HCl 100mM (8 pH at 37°C), MgCl2 50mM, KCl 1M, Triton™ X-100 0.2%, BSA 1mg/mL
Storage Requirements -20°C

OPTIZYME™ AP is a novel alkaline phosphatase that catalyzes the release of 5'- and 3'-phosphate groups from DNA, RNA and nucleotides in 10min.

Applications and Features:

  • OPTIZYME™ Alkaline Phosphatase dephosphorylates cloning vector DNA (10min at 37°C)
  • Fast thermoinactivation (5min at 75°C)
  • Simultaneous digestion and dephosphorylation of vector DNA
  • Single protocol for all types of DNA ends
  • Nucleotide degradation prior to sequencing PCR products
  • Active on nucleotides
  • Protein dephosphorylation
  • Active in all OPTIZYME restriction enzyme buffers

Supplied with:
10X OPTIZYME AP Buffer: 100mM Tris-HCl (pH 8.0 at 37°C), 50mM MgCl2, 1M KCl, 0.2% (v/v) Triton X-100, 1mg/mL BSA
Source: E. Coli cells with a cloned bacterial alkaline phosphatase gene
Quality Control:

  • Endo-, exodeoxyribonuclease and ribonuclease-free
  • Functionally tested for dephosphorylation of 5'-termini of overhanging, recessed and blunt DNA