Please login to your online account to display your discounted pricing

OPTIZYME™ Alkaline Phosphatase, Fisher BioReagents™


Manufacturer: fisher scientific company  BP80975

 View more versions of this product

Catalog No. BP80975

  • / Each
 

Description & Specifications

Specifications

Product Name Alkaline Phosphatase
Source E. coli cells with a cloned bacterial Alkaline Phosphatase gene
pH 7.4
Quantity 3000U
For Use With (Application) DePhosphorylates cloning vector DNA (10 min. at 37°C), fast thermo-inactivation (5 min. at 75°C), Nucleotide degradation prior to sequencing PCR products, active on Nucleotides, Protein dephosphorylation, restriction En
Storage Buffer OPTIZYME™ AP Buffer 10X : Tris HCl 100mM (8 pH at 37°C), MgCl2 50mM, KCl 1M, Triton™ X-100 0.2%, BSA 1mg/mL
Concentration 1U/μL
Color Colorless
Concentration 1U/μL
Storage Requirements -20°C
Format Solid
Quantity 3000U
Format Solid