OPTIZYME™ 10X Buffer BamHI, Fisher BioReagents™
Supplier:  Fisher BioReagents BP80891
Catalog No. BP8089-1
Description
Description
1X Buffer Composition:
10mM Tris-HCl (pH 8.0 at 37°C), 5mM MgCl2, 100mM KCl , 0.02 % Triton X-100, 0.1mg/mL BSA
Specifications
Specifications
10mM Tris HCl (pH 8 at 37°C), 5mM MgCl2, 100mM KCl, 0.02 % Triton™ X-100, 0.1mg/mL BSA
5 x 1 mL
Liquid
-20°C
Buffer BamHI