missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Opticap™ Sterile XL300 Capsule Filters
For sterilizing grade performance and mycoplasma removal
$484.73 - $590.93
Specifications
Sterility | Sterile |
---|---|
For Use With (Application) | Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals |
Length (English) | 4.7 in. |
Length (Metric) | 11.9 cm |
Catalog Number | Mfr. No. | Pore Size | Inlet Style | Outlet Style | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Pore Size | Inlet Style | Outlet Style | Price | Quantity | |||
KHVES003FF3
|
MilliporeSigma™
KHVES003FF3 |
0.5/0.1 μm | 0.75 in. (19mm)Sanitary Flange | 0.75 in. (19mm)Sanitary Flange |
Pack of 3 Each for $590.93
|
|
KHVES003FH3
|
MilliporeSigma™
KHVES003FH3 |
0.5/0.1 μm | 0.75 in. (19mm)Sanitary Flange | 0.56 in. (14mm) hose barb |
Pack of 3 Each for $590.93
|
|
KHGES003FF3
|
MilliporeSigma™
KHGES003FF3 |
0.5/0.2 μm | 0.75 in. (19mm)Sanitary Flange | 0.75 in. (19mm)Sanitary Flange |
Pack of 3 Each for $484.73
|
|
KHGES003HH3
|
MilliporeSigma™
KHGES003HH3 |
0.5/0.2 μm | 0.56 in. (14mm) Hose Barb | 0.56 in. (14mm) Hose Barb |
Pack of 3 Each for $484.73
|
|
KHGES003FH3
|
MilliporeSigma™
KHGES003FH3 |
0.5/0.2 μm | 0.75 in. (19mm)Sanitary Flange | 0.56 in. (14mm) Hose Barb |
Pack of 3 Each for $484.73
|
|
KHVES003HH3
|
MilliporeSigma™
KHVES003HH3 |
0.5/0.1 μm | 0.56 in. (14 mm) Hose Barb | 0.56 in. (14mm) Hose Barb |
Pack of 3 Each for $590.93
|
|
Description
- High-flux hydrophilic sterilizing-grade 0.1μm rated PES membrane for improved process economy
- On-board PES membrane prefilter for extended throughput in high fouling media
- Robust construction
- Withstand multiple sterilization cycles
- Easy wetting and integrity testing
- Broad chemical compatibility across a wide pH range (pH 1–14)
- Hydrophilic Polyethersulfone filter
- Polyester support
- Structural Components: Gamma stable polypropylene/Polysulfone
- Silicone vent O-rings
- Gamma compatible sterilization
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
Specifications
Sterile | |
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals | |
4.7 in. | |
11.9 cm |