missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MilliporeSigma™ Opticap™ Sterile XL300 Capsule Filters
For sterilizing grade performance and mycoplasma removal
Supplier: MilliporeSigma™ KHGES003FF3
Description
- High-flux hydrophilic sterilizing-grade 0.1μm rated PES membrane for improved process economy
- On-board PES membrane prefilter for extended throughput in high fouling media
- Robust construction
- Withstand multiple sterilization cycles
- Easy wetting and integrity testing
- Broad chemical compatibility across a wide pH range (pH 1–14)
- Hydrophilic Polyethersulfone filter
- Polyester support
- Structural Components: Gamma stable polypropylene/Polysulfone
- Silicone vent O-rings
- Gamma compatible sterilization
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals
Specifications
PES | |
Sterile | |
Cell culture media; Media additives; Serum; Concentrated protein; Process intermediates; Biologicals | |
0.75 in. (19mm)Sanitary Flange | |
11.9 cm |
0.5/0.2 μm | |
0.03 m2 | |
0.75 in. (19mm)Sanitary Flange | |
4.7 in. |