missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Optical Windows

Available in different materials and dimensions

Manufacturer:  Alfa Aesar™ 45885KS

 View more versions of this product

Catalog No. AA45885KS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Thickness: 2mm (0.08 in.)
25.4mm
Zinc selenide
1 in.
Surface finish: 60/40
Optical Window
Documents