missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Optical Windows

Available in different materials and dimensions

Manufacturer:  Alfa Aesar™ 45537KS

 View more versions of this product

Catalog No. AA45537KS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Thickness: 1mm (0.04 in.)
25.4mm
Quartz
1 in.
Surface finish: 40/20
Optical Window
Documents