Please login to your online account to display your discounted pricing
Optical Set
Complete with all necessary accessories to perform 36 experiments.
Manufacturer: fisher science education  HL2060-001
 View more versions of this product
Catalog No. S42208
  • / Each
Description & Specifications
Specifications
Product Type Light box and optical set
Includes Metal plate with single and triple slits, 12V line filament lamp, Lamp holder, Lucite™ prisms: three triangular, one rectangular and one semicircular, Lucite lenses: one biconcave, two biconvex, Mirrors: one each of plain, half cylindric
Quantity Each
Accessories
Accessories for Catalog Number: S42208
Item Mfr. No. Item Description Price Per Unit Quantity  
S96562
EISCO Low-Voltage Power Supply
eisco
PH1323
Compact regulated power supply. Each for $221.95