Please login to your online account to display your discounted pricing
Optical Set
Complete with all necessary accessories to perform 36 experiments.
Manufacturer: fisher scientific  C2060020
 View more versions of this product
Catalog No. S42580B
  • / Each
Description & Specifications
Specifications
Quantity Each
Product Type Raybox
  • Built-in, adjustable collimating lens
  • Provides a simple means of producing single or multiple light rays for use in elementary optics
  • Die-cast ventilated lamp housing, 5.9 x 2.75 x 2.55 in. (150 × 70 × 65mm) overall
  • Box has open front with finned sides to dissipate heat
  • Four apertures, one accommodates the slit plates to produce ray tracks, three for mounting a variety of color filters
  • Two side apertures fitted with hinged plane mirrors
  • Mirrors project beams that will overlap and merge with beam projected from fixed center aperture
  • Produces divergent, parallel, or convergent rays and allows for easy color mixing
  • Lenses, prisms, and mirrors are provided with handles to keep fingerprints off the optical faces
  • Light box will operate from any 12V, 3A, AC, or DC power supply
  • Power supply not included