Please login to your online account to display your discounted pricing
Optical Bench Set
Manufacturer: fisher science education  FC-400-U
 View more versions of this product
Catalog No. S423011
  • / Each
Description & Specifications
Specifications
Product Type Optical bench set
For Use With (Application) In a wide range of optics experiments including focal length of lenses, curvature or mirrors and lens combinations etc
Includes Aluminum Alloy bench, lamphouse with mounting rod, 2m of flexible cord with two 4mm plugs, Bulb (12V, 24W-B15), three universal holders on sliders, Triangular gauze object, Two pin objects, Diffuser or screen, Scale object, Magnification accessory, Double-sided screen, Plane mirror
Material Aluminum, Alloy bench
Quantity Each