United Scientific™ Optical Bench Set With Meter Stick
Optical Bench Set With Meter Stick contains sufficient equipment for high school physics applications.
$70.53 - $70.53
Specifications
Quantity | 1 Ea. |
---|---|
For Use With (Application) | High school physics applications |
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity | |||||
S05912
|
United Scientific™
OBSET1 |
Optical Bench Set |
Each for $70.53
|
|
|||||
Description
- Contents:
- 1 Meter Stick
- 1 Screen support
- 2 Lens Supports for 38 mm and 75 mm lenses/mirrors
- 5 Board screens measuring 10 x 12.5 cm with a millimeter scale on both sides
- 1 Marker and object
- 2 Metal supports for meter stick
- 1 Candle holder
- 1 Paraffin candle
Specifications
1 Ea. | |
High school physics applications |