Please login to your online account to display your discounted pricing

Optical Bench Set With Meter Stick


Contains sufficient equipment for high school physics applications.

Manufacturer: fisher science education  OBMS02

 View more versions of this product

Catalog No. S05921

  • / Each
 

Description & Specifications

Specifications

Product Type Metal support
For Use With (Application) High school physics applications
Quantity 2 Pack