Please login to your online account to display your discounted pricing

Optical Bench Set With Meter Stick

Contains sufficient equipment for high school physics applications.

Manufacturer: fisher science education  OBMRK1

 View all options for this product

Catalog No. S05916


Description & Specifications


Quantity Each
For Use With (Application) High school physics applications
Product Type Marker and object

  • Contents:
  • 1 Meter Stick
  • 1 Screen support
  • 2 Lens Supports for 38mm and 75mm lenses / mirrors
  • 5 Board screens measuring 10 x 12.5cm with a millimeter scale on both sides
  • 1 Marker and object
  • 2 Metal supports for meter stick
  • 1 Candle holder
  • 1 Paraffin candle