Please login to your online account to display your discounted pricing
Opti-Klens™ Wash Fountain
Chrome-plated, solid brass unit easily installs on any existing sink or basin faucet as a permanent eye wash fountain, yet does not interfere with normal faucet functioning.
Manufacturer: fisher science education  OPTIKLENSIEMD
 View more versions of this product
Catalog No. S96046A
  • / Each
Description & Specifications
Specifications
Product Type Opti-Klens I eyewash
For Use With (Application) Eye/facewash
Includes Diverter, Chrome Plated Brass valve, adapters, dust cover
Certifications/Compliance ANSI Z358.1-1990