Opti-Klens™ Wash Fountain
Chrome-plated, solid brass unit easily installs on any existing sink or basin faucet as a permanent eye wash fountain, yet does not interfere with normal faucet functioning.
$179.55 - $179.55
Specifications
Certifications/Compliance ANSI Z358.1-1990
For Use With (Application) Eye/facewash
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Includes Product Type Price Quantity  
Catalog Number Mfr. No. Includes Product Type Price Quantity  
S96046A Fisher Science Education
OPTIKLENSIEMD
Diverter, Chrome Plated Brass valve, adapters, dust cover Opti-Klens I eyewash Each for $179.55
Specifications
Description
  • Water supply in an emergency is limitless, unlike with portable models
  • Unit frees both hands to hold eyes open to the flow of water
  • No new plumbing is required
  • Supplied with two of the most common adapters
  • Other adapters are available from the manufacturer at no additional cost
  • Spray configuration on Opti-Klens II allows both eyes and face to be washed
Compliance
Meets ANSI Standard #Z359.1-1990 for an eye wash unit.
Description & Specifications
Specifications
ANSI Z358.1-1990
Eye/facewash