Please login to your online account to display your discounted pricing
Thermo Scientific™ Open-top Caps without Septum for 8-425 Standard Opening Vials
Thermo Scientific™ National Open-Top Caps without Septum for 8-425 Standard Opening Vials are designed to be used with any 8mm septa.
Manufacturer: thermo scientific™  C401398R
 View more versions of this product
 Disclaimers
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Catalog No. 03-378-336
  • / Pack of 100 Each
Description & Specifications
Specifications
Description PP; Red; Flanged for Shimadzu
Closure Color Red
Closure Material Polypropylene
Vial Closure Size 8-425mm
Type Open top cap with flange
For Use With 8-425 Standard Opening Vials
Item Description PP; Red; Flanged for Shimadzu
Thermo Scientific™ National Open-Top Caps without Septum for 8-425 Standard Opening Vials are designed to be used with any 8mm septa.
  • Open top caps for use with any 8mm septa
  • Phenolic caps are suitable for autoclaving and low temperature applications
  • Polypropylene caps are chemically inert and suitable for most chromatography applications
  • Flanged caps are particularly suitable for many Japanese autosamplers
  • Kombo-Kit contains 200 each of black, blue, green, red, and yellow flanged caps