Please login to your online account to display your discounted pricing

OPC 21268, Tocris™

Orally active vasopressin V1 antagonist

$290.95 - $1,205.80

Chemical Identifiers

Chemical Name or Material OPC 21268
Assay Percent Range >98%
Molecular Formula C26H31N3O4
Formula Weight 449.54
Synonym N-[3-[4-[[4-(3, 4-Dihydro-2-oxo-1(2H)quinolinyl)-1-piperidinyl]carbonyl]phenoxy]propyl]-acetamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

392450

 
r & d systems
RDS392450
50mg Each for $1,205.80

392410

 
r & d systems
RDS392410
10mg Each for $290.95
Description & Specifications

Specifications

Formula Weight 449.54
Quantity 50mg
Synonym N-[3-[4-[[4-(3, 4-Dihydro-2-oxo-1(2H)quinolinyl)-1-piperidinyl]carbonyl]phenoxy]propyl]-acetamide
Chemical Name or Material OPC 21268
Assay Percent Range >98%
Molecular Formula C26H31N3O4
CAS 131631-89-5
Recommended Storage Store at +4°C
Safety and Handling

Store at +4°C