missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dunlop™ Onguard™ Women's White PVC Boots

$49.00 - $49.00

Specifications

Certifications/Compliance ASTM F2413-05
Color White
Material PVC
For Use With (Application) Used in commercial fishing, shrimping, and food processing
Height (English) 14 in.
View More Specs

Products 3
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
29-635-9123
View Documents
Dunlop™
510340500
5
Pair for $49.00
 
29-635-9124
View Documents
Dunlop™
510340600
6
Pair for $49.00
 
29-635-9126
View Documents
Dunlop™
510340800
8
Pair for $49.00
 
Description

Description

Specifications

Specifications

ASTM F2413-05
PVC
14 in.
Mild chemicals
Safety-Loc™ outsole and replaceable cushion insoles
White
Used in commercial fishing, shrimping, and food processing
Kick-off
Seamless
SDS