Dunlop™ Onguard™ White PVC Boots
Manufacturer:  Dunlop™ 810120800
 View more versions of this product
Catalog No. 29-635-9131
Description
Description
Specifications
Specifications
Chemicals
White
Used in commercial fishing, shrimping, and food processing
Kick-off
Steel
White Safety-Loc™ outsole and replaceable cushion insoles, slip-resistant
Steel Toe
ASTM F2413-05
Pull-on
16 in.
PVC
8
Steel