Please login to your online account to display your discounted pricing

Onguard™ Superpoly Boots with Chevron Sole

Ideal for use in food processing, especially red-meat processing and petrochemical applications

$60.80 - $60.80

Specifications

Certifications/Compliance ASTM F2413-05, ANSI Z41 PT99 M I/75 C/75 PR
Closure Type Pull-up
Color Brick red
Material Urethane, PVC
For Use With (Application) Used in slaughter houses, red meat processing, canneries, breweries, dairies, oil rigs and chemical plants
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-024361

 
onguard industries
853240700
7 Pack for $60.80
Case of 6 Pack for $343.68

19-024355

 
onguard industries
853240800
8 Pack for $60.80
Case of 6 Pack for $343.68

19-024357

 
onguard industries
853241100
11 Pack for $60.80
Case of 6 Pack for $343.68

19-024354

 
onguard industries
853240900
9 Pack for $60.80
Case of 6 Pack for $343.68

19-024360

 
onguard industries
853241200
12 Pack for $60.80
Case of 6 Pack for $343.68

19-074059

 
onguard industries
853240500
5 Pack for $60.80
Case of 6 Pack for $343.68
Description & Specifications

Specifications

Certifications/Compliance ASTM F2413-05, ANSI Z41 PT99 M I/75 C/75 PR
Closure Type Pull-up
Color Brick red
Material Urethane, PVC
For Use With (Application) Used in slaughter houses, red meat processing, canneries, breweries, dairies, oil rigs and chemical plants
Gender Male
Height (English) 16 in.
Lug Type Kick off
Resistant Chemical
Shank Material Steel
Sole Features Tan Chevron outsole and replaceable cushion insoles
Toe Material Steel

  • Waterproof with a high degree of chemical resistance
  • Steel toe and shank provide optimal protection
  • Chevron outsole provides excellent slip resistance
  • Pull-up and kick-off lugs
  • Removable or replaceable preformed insole for added comfort

Steel toe meets or exceeds ASTM™ F2413-11 specifications