missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Dunlop™ Onguard™ Steel Midsole Work Boots

Ideal for demolition crews and recycling plants

$96.58 - $96.58

Specifications

Certifications/Compliance ASTM
Color Black
Material Polyblend
For Use With (Application) Demolition crews and recycling plants
Height (English) 16 in.
View More Specs

Products 8
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
19-024512
View Documents
Dunlop™
868020700
7
Pack for $96.58
Case of 6 Pack for $545.88
 
19-024513
View Documents
Dunlop™
868021000
10
Pack for $96.58
Case of 6 Pack for $545.88
 
19-024509
View Documents
Dunlop™
868020600
6
Pack for $96.58
Case of 6 Pack for $545.88
 
19-024511
View Documents
Dunlop™
868020900
9
Pack for $96.58
Case of 6 Pack for $545.88
 
19-024514
View Documents
Dunlop™
868021100
11
Pack for $96.58
Case of 6 Pack for $545.88
 
19-024515
View Documents
Dunlop™
868021200
12
Pack for $96.58
Case of 6 Pack for $545.88
 
19-024516
View Documents
Dunlop™
868021300
13
Pack for $96.58
Case of 6 Pack for $545.88
 
19-080644
View Documents
Dunlop™
868020800
8
Pack for $96.58
Case of 6 Pack for $545.88
 
Description

Description

  • Polyblend PVC construction provides moderate chemical protection
  • Full steel midsole for complete underfoot protection
  • Steel toe for optimal impact and crush resistance
  • Feature Ultragrip™ sole, pull-up and kick-off lugs, and replaceable insoles
  • 16 in. high (41cm)
  • Waterproof
  • Black

Compliance

Stainless-steel midsole meets or exceed ASTM™ F2413-11 for puncture resistance and impact and crush resistance. Steel midsole and toe also meet CSA Z195 Grade 1.

Specifications

Specifications

ASTM
Polyblend
16 in.
Standard
Black
Demolition crews and recycling plants
Ultragrip™ sole, full Steel midsole
Steel
SDS