missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dunlop™ Onguard™ Pullover Slicker Boots
Fit easily over work shoes
$56.70 - $56.91
Specifications
Buckle Material | Plastic |
---|---|
Color | Yellow |
Material | Flexothane,PVC |
For Use With (Application) | Construction, concrete work, utilities and food processing |
Height (English) | 17 in. |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
29-635-2198
|
Dunlop™
880501200 |
12 |
Pair for $56.70
|
|
19-024522
|
Dunlop™
880501000 |
10 |
Pair for $56.91
Case of 6 Pair for $341.46 |
|
19-024523
|
Dunlop™
880501100 |
11 |
Pack for $56.91
Case of 6 Pack for $341.46 |
|
19-024525
|
Dunlop™
880501400 |
14 |
Pack for $56.91
Case of 6 Pack for $341.46 |
|
19-024527
|
Dunlop™
880500800 |
8 |
Pack for $56.91
Case of 6 Pack for $341.46 |
|
19-024526
|
Dunlop™
880500700 |
7 |
Pair for $56.91
Case of 6 Pair for $341.46 |
|
19-024528
|
Dunlop™
880500900 |
9 |
Pair for $56.91
|
|
19-024529
|
Dunlop™
880501300 |
13 |
Pack for $56.91
Case of 6 Pack for $341.46 |
|
Description
- Waterproof with excellent cold weather flexibility and chemical resistance
- Nonconductive adjustable polyester strap with plastic buckle and snaps
- Nonabsorbent polyester lining
- 17 in. high (43cm)
- Yellow with black cleated soles and kick-off lugs
Specifications
Plastic | |
Flexothane,PVC | |
17 in. | |
Black self-cleaning cleated outsole |
Yellow | |
Construction, concrete work, utilities and food processing | |
Polyester | |
Polyester |