Please login to your online account to display your discounted pricing

Onguard Protex Bib Overall

$20.36 - $20.36

Specifications

Ankle Style Take up snap
Closure Type Plain
Color Yellow
Material PVC, Nylon, Polyester
No. of Pockets 2.0
View More Specs
Products
Catalog Number Mfr. No. Length (English) Inseam Length (English) Size Size (English) Waist Price Quantity    

19-806-570

 
onguard industries
79050
32 in. 56.5 in. 2X-Large 58 in. Each for $20.36
Description & Specifications

Specifications

Ankle Style Take up snap
Closure Type Plain
Color Yellow
Material PVC, Nylon, Polyester
No. of Pockets 2.0
Style Protex™
Suspender Adjustable elastic
For Use With (Application) Heavy duty protection for every Industry
Thickness 0.3mm
Waist Style Adjustable snap