Please login to your online account to display your discounted pricing

Onguard Men's Steel Toe Comfort NeoMax Boots with Tunnel Outsole

$98.69 - $98.69

Specifications

Certifications/Compliance ASTM F2413-05
Color Grey
Material PVC
For Use With (Application) Used in variety of applications
Gender Male
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

29-663-813

 
onguard industries
820021000
10 Pair for $98.69
Case of 6 Pair for $557.82
Description & Specifications

Specifications

Certifications/Compliance ASTM F2413-05
Color Grey
Material PVC
For Use With (Application) Used in variety of applications
Gender Male
Height (English) 15 in.
Shank Material Steel
Sole Features Tunnel outsole and abrasion resistant outsole, slip resistant
Toe Material Steel