Please login to your online account to display your discounted pricing

Onguard Men's Slicker Overboots with Self-Cleaning Cleated Outsole

$42.28 - $42.28

Specifications

Buckle Material Plastic
Color Black
Material PVC, Flex-O-Thane
For Use With (Application) Construction, concrete work, utilities and food processing
Gender Male
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-051945

 
onguard industries
860501200
12 Pack for $42.28

19-051947

 
onguard industries
860501300
13 Pack for $42.28
Description & Specifications

Specifications

Buckle Material Plastic
Color Black
Material PVC, Flex-O-Thane
For Use With (Application) Construction, concrete work, utilities and food processing
Gender Male
Height (English) 17 in.
Liner Material Polyester
Sole Features Self-cleaning cleated outsole
Strap Material Polyester