Please login to your online account to display your discounted pricing

Onguard Industries DuraPro™ XCP Chemical Resistant Plain Toe Boots with Steel Shank

High performance compound provides ideal chemical resistance in the upper and superior abrasion and slip resistance in the outsole

$59.54 - $59.54

Specifications

Sole Features Next Generation Chevron, slip resistant
Gender Male
Shank Material Steel
Toe Style Plain
Color Brown
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

18-500-528

 
onguard industries
840853
3 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-529

 
onguard industries
840854
4 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-530

 
onguard industries
840855
5 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-531

 
onguard industries
840856
5 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-532

 
onguard industries
840857
7 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-533

 
onguard industries
840858
8 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-534

 
onguard industries
840859
9 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-535

 
onguard industries
8408510
10 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-536

 
onguard industries
8408511
11 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-537

 
onguard industries
8408512
12 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-538

 
onguard industries
8408513
13 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-539

 
onguard industries
8408514
14 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-540

 
onguard industries
8408515
15 Pair for $59.54
Pack of 6 Pair for $336.54

18-500-541

 
onguard industries
8408516
16 Pair for $59.54
Pack of 6 Pair for $336.54
Description & Specifications

Specifications

Sole Features Next Generation Chevron, slip resistant
Gender Male
Shank Material Steel
Toe Style Plain
Color Brown
Resistant Chemicals
Material Superpoly Upper; Sureflex Outsole

  • Sloped comfort top rim prevents chafing and reduces discomfort when walking, bending and kneeling
  • 360° pull tab for easy donning
  • Steel shank protects and supports arch
  • Larger top circumference for quick and easy pull-on
  • 3/4 in. height inner liner allows  increased flexibility, subtle feel at the calf, and soft lined feel of leg and foot
  • Honeycomb design molded inside the heel to reduce impact when walking
  • Ergonomically designed foot-bed to provide better underfoot support
  • Protective bumpers at rear of boot to prevent premature wear/puncture
  • Extended circumference kick-off ledge
  • Composition: Superpoly Upper; Sureflex Outsole
  • Sole Design: Next Generation Chevron
  • Available in sizes ranging from 3 to 16