Dunlop™ Onguard™ Hazmax™ EZ-Fit™ Boots
Designed for easy donning and doffing while wearing chemical protective suit
Manufacturer:  Dunlop™ 87015M
 View more versions of this product
Catalog No. 29-635-4054
Alert:
Caution! Do not use for fire or electrical hazard protection.
Description
Description
  • One-piece injection molded with no buttons or elastic bands to break for easier decontamination
  • Large kick-off lug for easy removal
  • Wider across the instep for easy entry
  • Nonslip EZ decon outsole for easier decontamination and cleanup
  • Steel toe, steel shank, and steel midsole for complete protection
  • Height: 11 in. (28cm)
Order Info
Order normal shoe size–designed to accommodate a fully encapsulated suit. Men's full sizes: Small: 6-8, Medium: 9-10, Large: 11-12, X-Large: 13-15.
Certifications
Certified by S.E.I. to meet the requirements of the NFPA 1991, 2005 Edition Standard on Vapor-Protective ensembles for Hazardous Materials Emergencies.
Compliance
Meets ASTM™ F2413-11; CSA Z195 requirements. NFPA 1991 Standard on Vapor-Protective Ensembles for Hazardous Materials Emergencies and CBRN Terrorism Incidents, 2016.
Specifications
Specifications
For situation involving dangerous chemicals, gases and waste
11 in.
28 cm
Medium