Dunlop™ Onguard™ Hazmax™ Boots
Choose from two styles
Manufacturer:  Dunlop™ 870120800
 View more versions of this product
Catalog No. 17-985-403D
Alert:
Do not use for fire protection.
Description
Description
Ideal for emergency response teams, hazmat technicians, environmental specialists, and site remediation workers.
Over-the-sock knee boots
  • Steel toes
  • Full steel midsoles and steel ladder shanks
  • Limited flash fire protection; for escape only in the event of a chemical fire
  • Preformed three-layer comfort insoles
  • Kick-off lug
  • Green chemical-resistant uppers
  • Yellow Ultragrip™ outsoles
  • No snaps or buttons for more reliable fit
Overboot
  • Adjustable nonconductive polyester straps
  • Nonmetallic buckles and snaps
  • Kick-off lug
  • Black cleated outsoles
Order Info
When ordering over-the-sock boots to be worn over a chemical protective suit with attached booties, order two shoe sizes larger. When ordering overboots, order regular shoe size. Results of chemical permeation tests available by calling Fisher Safety Customer Service at 1-800-772-6733.
Compliance
Meets ASTM™ F2413-05; CSA Z195 requirements.
Specifications
Specifications
Safety
Ideal for emergency response teams, hazmat technicians, environmental specialists, and site remediation workers.
Flexible PVC alloy
Yellow
16 in.
8
Documents