Please login to your online account to display your discounted pricing

Onguard™ Economy General-Purpose Knee Boots

Made from 90% recycled PVC plastic

$22.58 - $30.02

Specifications

Certifications/Compliance ASTM F2413-11, ANSI Z41 PT99 M I/75 C/75
Closure Type Pull-on
Color Black
Material PVC
For Use With (Application) Used in orchards, vegetable fields, citrus groves, and agriculture
View More Specs
Products
Catalog Number Mfr. No. Toe Style Size Price Quantity    

17986388

 
onguard industries
866050700*
Plain 7 Pair for $23.25

17986387

 
onguard industries
866050600*
Plain 6 Pair for $23.25

17986389

 
onguard industries
866050800*
Plain 8 Pair for $23.25

17986391

 
onguard industries
866051000*
Plain 10 Pair for $23.25

17986392

 
onguard industries
866051100*
Plain 11 Pair for $23.25

17986393

 
onguard industries
866051200*
Plain 12 Pair for $23.25

17986394

 
onguard industries
866051300*
Plain 13 Pair for $23.25

17986390

 
onguard industries
866050900*
Plain 9 Pair for $23.25

29-635-6277

 
onguard industries
866060600
Steel 6 Pair for $30.02

27-058-00

 
onguard industries
866060700
Steel 7 Pair for $30.02

27-058-01

 
onguard industries
866060800
Steel 8 Pair for $30.02

27-058-02

 
onguard industries
866060900
Steel 9 Pair for $29.78

27-058-04

 
onguard industries
866061100
Steel 11 Pair for $30.02

27-058-05

 
onguard industries
866061200
Steel 12 Pair for $30.02

27-058-03

 
onguard industries
866061000
Steel 10 Pair for $29.16

29-635-6269

 
onguard industries
866050600
Plain 6 Pair for $23.25

29-635-6270

 
onguard industries
866050700
Plain 7 Pair for $22.58

29-635-6271

 
onguard industries
866050800
Plain 8 Pair for $22.58

29-635-6272

 
onguard industries
866050900
Plain 9 Pair for $23.47

29-635-6273

 
onguard industries
866051000
Plain 10 Pair for $25.85

29-635-6274

 
onguard industries
866051100
Plain 11 Pair for $22.58

29-635-6275

 
onguard industries
866051200
Plain 12 Pair for $23.47

29-635-6276

 
onguard industries
866051300
Plain 13 Pair for $23.25
Description & Specifications

Specifications

Certifications/Compliance ASTM F2413-11, ANSI Z41 PT99 M I/75 C/75
Closure Type Pull-on
Color Black
Material PVC
For Use With (Application) Used in orchards, vegetable fields, citrus groves, and agriculture
Gender Male
Height (English) 16 in.
Lug Type Kick off
Sole Features Cleated outsole and replaceable cushion insoles
Waterproof Yes

Over-the-sock boots for wet or muddy conditions only—not for chemical or electrical resistance. Excellent for a wide range of industrial and agricultural applications.

  • Cleated outsoles, replaceable insoles, and kick-off lugs
  • Cut boot at cut-off line to shorten
  • Replaceable insole for greater comfort
  • Height: 16 in. (41cm)
  • Black
  • Environmentally Friendly: Item contains environmental attributes or features (e.g. water conserving, lead-free, etc.) as described in the item detail
  • Recycled: Item contains recycled content as described in the item detail

Available with or without steel toe.

Steel-toe model meets or exceeds ASTM™ F2413-11.