Please login to your online account to display your discounted pricing

Onguard™ Buckle Arctic Boots

Waterproof and durable rubber

$47.00 - $47.00

Specifications

Color Black
Material Rubber
For Use With (Application) Construction, utilities, roadwork and foul weather
Height (English) 11 in.
Liner Material Cotton
View More Specs
Products
Catalog Number Mfr. No. Size Price Quantity    

19-024388

 
onguard industries
860631400
14 Pack for $47.00
Case of 6 Pack for $265.68

19-024389

 
onguard industries
860631500
15 Pack for $47.00
Case of 6 Pack for $265.68

19-024391

 
onguard industries
860631100
11 Pack for $47.00
Case of 6 Pack for $265.68

19-024392

 
onguard industries
860631200
12 Pack for $47.00
Case of 6 Pack for $265.68

19-024390

 
onguard industries
860631000
10 Pack for $47.00
Case of 6 Pack for $265.68

19-024393

 
onguard industries
860631300
13 Pack for $47.00
Case of 6 Pack for $265.68

19-024394

 
onguard industries
860630700
7 Pack for $47.00
Case of 6 Pack for $265.68

19-024396

 
onguard industries
860630900
9 Pack for $47.00
Case of 6 Pack for $265.68

19-024395

 
onguard industries
860630800
8 Pack for $47.00
Case of 6 Pack for $265.68
Description & Specifications

Specifications

Color Black
Material Rubber
For Use With (Application) Construction, utilities, roadwork and foul weather
Height (English) 11 in.
Liner Material Cotton
Sole Features Stretch net lining, self cleaning, self draining gusset, four noncorrosive buckles, cleated soles

  • Provide maximum protection against the elements
  • 11 in. high (28cm) boots fit over most work shoes and boots
  • Feature cleated soles, four noncorrosive buckles, stretch-net lining inside and self-draining gusset