missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Dunlop™ Onguard™ Blue Max™ Work Boots
Moderate chemical resistance for work in poultry and food processing
Manufacturer: Dunlop™ 891021300
Description
- Extremely abrasion resistant with excellent traction and slip resistance
- Ultragrip™ sole, steel shank
- Height: 16 in. (41cm)
- Waterproof
- Blue
Order Info
Available with plain or steel toe.
Compliance
Steel toe meets or exceeds ASTM™ F2413-11 specifications
Specifications
ASTM | |
Excellent for use in poultry, seafood processing, food processing and canneries | |
PVC | |
13 | |
Steel |
Blue | |
16 in. | |
Steel | |
Replaceable cushion insoles, slip resistance, Ultragrip™ sipe outsole | |
Steel Toe |