Please login to your online account to display your discounted pricing

Onguard™ Blue Max™ Work Boots

Moderate chemical resistance for work in poultry and food processing

$67.18 - $67.18

Specifications

Certifications/Compliance ASTM
Color Blue
Material PVC
For Use With (Application) Excellent for use in poultry, seafood processing, food processing and canneries
Gender Male
View More Specs
Products
Catalog Number Mfr. No. Toe Style Size Price Quantity    

19-068957

 
onguard industries
891021100
Steel 11 Pack for $67.18
Case of 6 Pack for $379.74

19-068987

 
onguard industries
891021000
Steel 10 Pack for $67.18
Case of 6 Pack for $379.74

19-068986

 
onguard industries
891020900
Steel 9 Pack for $67.18
Case of 6 Pack for $379.74

19-069000

 
onguard industries
891021200
Steel 12 Pack for $67.18
Case of 6 Pack for $379.74

19-069002

 
onguard industries
891021300
Steel 13 Pack for $67.18
Case of 6 Pack for $379.74
Description & Specifications

Specifications

Certifications/Compliance ASTM
Color Blue
Material PVC
For Use With (Application) Excellent for use in poultry, seafood processing, food processing and canneries
Gender Male
Height (English) 16 in.
Shank Material Steel
Sole Features Replaceable cushion insoles, slip resistance, Ultragrip™ sipe outsole

  • Extremely abrasion resistant with excellent traction and slip resistance
  • Ultragrip™ sole, steel shank
  • Height: 16 in. (41cm)
  • Waterproof
  • Blue

Available with plain or steel toe.

Steel toe meets or exceeds ASTM™ F2413-11 specifications