One-Piece TFE Stoppers for Ground Joints
$84.91 - $84.91
Catalog Number | Mfr. No. | Standard Taper Joint | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Standard Taper Joint | Price | Quantity | |||||
14-645-20G
|
Scientific Machine
82216 |
24/25 |
Each for $84.91
|
|
Description
- Hollow cup allows use for microanalytical work when inverted
- Unaffected by corrosive chemicals; unbreakable
- Need no lubricants
- Snug fit, easy removal
- Weight of top keeps hollow tip up (all stoppers are hollow except 14-645-35B), prevents contamination of wetted surface when placed on lab bench
- Use to replace ground joint glass stoppers
Specifications
24/25 |