Scientific Machine One-Piece TFE Stoppers for Ground Joints
Manufacturer:  Scientific Machine 82216
 View more versions of this product
Catalog No. 14-645-20G
Description
Description
  • Hollow cup allows use for microanalytical work when inverted
  • Unaffected by corrosive chemicals; unbreakable
  • Need no lubricants
  • Snug fit, easy removal
  • Weight of top keeps hollow tip up (all stoppers are hollow except 14-645-35B), prevents contamination of wetted surface when placed on lab bench
  • Use to replace ground joint glass stoppers
Specifications
Specifications
24/25
PTFE