SciMac One-Piece TFE Stoppers
PTFE-resin stoppers will not freeze or bind, chemically or mechanically
$55.06 - $85.26
Catalog Number | Mfr. No. | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Price | Quantity | ||||||
14-645-35D
|
SciMac
84416* |
Each for $55.06
|
|
14-645-35G
|
SciMac
84427 |
Each for $85.26
|
|
Description
- Hollow cup allows use for microanalytical work when inverted
- Unaffected by corrosive chemicals; unbreakable
- Need no lubricants
- Snug fit, easy removal
- Weight of top keeps hollow tip up (all stoppers are hollow except 14-645-35B, 14-645-20C and 14-645-20D), prevents contamination of wetted surface when placed on lab bench
Specifications
PTFE |