SciMac One-Piece TFE Stoppers
PTFE-resin stoppers will not freeze or bind, chemically or mechanically
Manufacturer:  SciMac 84427
 View more versions of this product
Catalog No. 14-645-35G
Description
Description
  • Hollow cup allows use for microanalytical work when inverted
  • Unaffected by corrosive chemicals; unbreakable
  • Need no lubricants
  • Snug fit, easy removal
  • Weight of top keeps hollow tip up (all stoppers are hollow except 14-645-35B, 14-645-20C and 14-645-20D), prevents contamination of wetted surface when placed on lab bench
Specifications
Specifications
PTFE
27