Please login to your online account to display your discounted pricing

Ondansetron hydrochloride, Tocris

Selective 5-HT3 antagonist

$139.08 - $580.33

Chemical Identifiers

Chemical Name or Material Ondansetron hydrochloride
Assay Percent Range >99%
Molecular Formula C18H19N3O·HCl
Formula Weight 329.83
Synonym 1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

289150

 
r & d systems
RDS289150
50mg Each for $580.33

289110

 
r & d systems
RDS289110
10mg Each for $139.08
Description & Specifications

Specifications

Formula Weight 329.83
Quantity 50mg
CAS 99614-01-4
Chemical Name or Material Ondansetron hydrochloride
Assay Percent Range >99%
Molecular Formula C18H19N3O·HCl
Synonym 1,2,3,9-Tetrahydro-9-methyl-3-[(2-methyl-1H-imidazol-1-yl)methyl]-4H-carbazol-4-one hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature