missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ondansetron Hydrochloride Dihydrate, TCI America™

Manufacturer:  TCI America O04075G

 View more versions of this product

Catalog No. O04075G



Specifications

Specifications

Ondansetron Hydrochloride Dihydrate
178°C
103639-04-9
MFCD00374371
1,2,3,9-tetrahydro-9-methyl-3-2-methyl-1h-imidazol-1-yl methyl-4h-carbazol-4-one hydrochloride; gr 38032f; ondansetron hydrochloride
CC1=NC=CN1CC2CCC3=C(C2=O)C4=CC=CC=C4N3C.O.O.[Cl]
364.85
329.83
≥98.0% (HPLC,T)
White-Yellow
5g
C18H23ClN3O3
2811
BGGIFKYQGVGSIW-UHFFFAOYSA-N
chlorine;9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one;dihydrate
71317260
Crystal-Powder at 20°C

Chemical Identifiers

103639-04-9
364.85
BGGIFKYQGVGSIW-UHFFFAOYSA-N
71317260
CC1=NC=CN1CC2CCC3=C(C2=O)C4=CC=CC=C4N3C.O.O.[Cl]
C18H23ClN3O3
MFCD00374371
1,2,3,9-tetrahydro-9-methyl-3-2-methyl-1h-imidazol-1-yl methyl-4h-carbazol-4-one hydrochloride; gr 38032f; ondansetron hydrochloride
chlorine;9-methyl-3-[(2-methylimidazol-1-yl)methyl]-2,3-dihydro-1H-carbazol-4-one;dihydrate
Safety and Handling

Safety and Handling

RTECSNumber : FE6375500

TSCA : No

Recommended Storage : Refrigerator

Documents