missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oncrasin 1, TCI America™

Manufacturer:  TCI America O045050MG

 View more versions of this product

Catalog No. O045050MG



Specifications

Specifications

Oncrasin 1
117°C
75629-57-1
MFCD01051808
ZDRQMXCSSAPUMM-UHFFFAOYSA-N
1-[(4-chlorophenyl)methyl]indole-3-carbaldehyde
872445
Crystal-Powder at 20°C
Undesignated
50mg
C16H12ClNO
oncrasin-1, 1-4-chlorobenzyl-1h-indole-3-carbaldehyde, oncrasin 1, 1-4-chlorophenyl methyl indole-3-carbaldehyde, 1-4-chlorophenyl methyl-1h-indole-3-carboxaldehyde, 1-4-chloro-benzyl-1h-indole-3-carbaldehyde, oncrasin1, cambridge id 5601604, n-4-chlorobenzyl-3-formylindole, oncrasin hplc
C1=CC=C2C(=C1)C(=CN2CC3=CC=C(C=C3)Cl)C=O
269.728
269.73
≥95.0% (HPLC)

Chemical Identifiers

75629-57-1
269.728
ZDRQMXCSSAPUMM-UHFFFAOYSA-N
872445
C1=CC=C2C(=C1)C(=CN2CC3=CC=C(C=C3)Cl)C=O
C16H12ClNO
MFCD01051808
oncrasin-1, 1-4-chlorobenzyl-1h-indole-3-carbaldehyde, oncrasin 1, 1-4-chlorophenyl methyl indole-3-carbaldehyde, 1-4-chlorophenyl methyl-1h-indole-3-carboxaldehyde, 1-4-chloro-benzyl-1h-indole-3-carbaldehyde, oncrasin1, cambridge id 5601604, n-4-chlorobenzyl-3-formylindole, oncrasin hplc
1-[(4-chlorophenyl)methyl]indole-3-carbaldehyde
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents