missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Oncrasin 1 95.0+%, TCI America™

$200.14 - $704.31

Chemical Identifiers

CAS 75629-57-1
Molecular Formula C16H12ClNO
Molecular Weight (g/mol) 269.728
MDL Number MFCD01051808
InChI Key ZDRQMXCSSAPUMM-UHFFFAOYSA-N
Synonym oncrasin-1, 1-4-chlorobenzyl-1h-indole-3-carbaldehyde, oncrasin 1, 1-4-chlorophenyl methyl indole-3-carbaldehyde, 1-4-chlorophenyl methyl-1h-indole-3-carboxaldehyde, 1-4-chloro-benzyl-1h-indole-3-carbaldehyde, oncrasin1, cambridge id 5601604, n-4-chlorobenzyl-3-formylindole, oncrasin hplc
PubChem CID 872445
IUPAC Name 1-[(4-chlorophenyl)methyl]indole-3-carbaldehyde
SMILES C1=CC=C2C(=C1)C(=CN2CC3=CC=C(C=C3)Cl)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
O045010MG View Documents TCI America
O045010MG
10mg Each for $200.14
 
O045050MG View Documents TCI America
O045050MG
50mg Each for $704.31
 
Specifications

Chemical Identifiers

75629-57-1
269.728
ZDRQMXCSSAPUMM-UHFFFAOYSA-N
872445
C1=CC=C2C(=C1)C(=CN2CC3=CC=C(C=C3)Cl)C=O
C16H12ClNO
MFCD01051808
oncrasin-1, 1-4-chlorobenzyl-1h-indole-3-carbaldehyde, oncrasin 1, 1-4-chlorophenyl methyl indole-3-carbaldehyde, 1-4-chlorophenyl methyl-1h-indole-3-carboxaldehyde, 1-4-chloro-benzyl-1h-indole-3-carbaldehyde, oncrasin1, cambridge id 5601604, n-4-chlorobenzyl-3-formylindole, oncrasin hplc
1-[(4-chlorophenyl)methyl]indole-3-carbaldehyde

Specifications

Undesignated
10mg
C16H12ClNO
oncrasin-1, 1-4-chlorobenzyl-1h-indole-3-carbaldehyde, oncrasin 1, 1-4-chlorophenyl methyl indole-3-carbaldehyde, 1-4-chlorophenyl methyl-1h-indole-3-carboxaldehyde, 1-4-chloro-benzyl-1h-indole-3-carbaldehyde, oncrasin1, cambridge id 5601604, n-4-chlorobenzyl-3-formylindole, oncrasin hplc
C1=CC=C2C(=C1)C(=CN2CC3=CC=C(C=C3)Cl)C=O
269.728
269.73
≥95.0% (HPLC)
117°C
75629-57-1
MFCD01051808
ZDRQMXCSSAPUMM-UHFFFAOYSA-N
1-[(4-chlorophenyl)methyl]indole-3-carbaldehyde
872445
Crystal-Powder at 20°C
Oncrasin 1
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator