Please login to your online account to display your discounted pricing
Thermo Scientific™ Omnistrips
Achieve consistent PCR results using uniform, thin walled tubes with low binding interior surface ensuring maximum yield
Manufacturer: thermo scientific™  AB0485
 View more versions of this product
Catalog No. AB-0485
  • / Case of 250 Each
Description & Specifications
Specifications
Description Omnistrip tubes without caps
Certifications/Compliance Certified free of human DNA, RNase, DNase, and endotoxins.
Type Tubes
Item Description Omnistrip Tubes, No Caps
The Thermo Scientific Omnistrips feature uniform, thin walls that give optimal and consistent heat transfer, reducing variation between samples. The low binding interior surface prevents reaction components from binding to side walls, ensuring maximum yield. The tight sealing strip caps significantly reduce evaporation to generate more efficient and higher yield results.
Highlights
  • Suitable for 0.5 mL thermal cycler blocks
  • 0.32 mL maximum tube capacity when capped
  • 8-tube strip
  • Sealable using Flat and Domed Cap Strips
  • Also available in a pre-assembled 96-well Omni-Tube Plate
Applications
  • PCR