Please login to your online account to display your discounted pricing

Omnifit Tube End Fittings


1/4 in.-28 UNF male threaded polypropylene fittings

Manufacturer: bio chem fluidics inc  002110

 View more versions of this product

Catalog No. OF002110

  • / Pack of 10 Each
 

Description & Specifications

Specifications

Type Tube End Fitting
Material Polypropylene
Color Assorted
For Use With 1/16 in. O.D. Tubing