Please login to your online account to display your discounted pricing
Omnifit Tube End Fittings
1/4 in.-28 UNF male threaded polypropylene fittings
$54.70 - $54.70
Products
Catalog Number Mfr. No. Price Quantity    
OF002110
bio chem fluidics inc
002110
Pack of 10 Each for $54.70
Description & Specifications
  • Won't compress tubing; allow it to be free spinning
  • Must be used with Gripper Fittings
.